Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXLNA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018057634.1 | XP_017913123.1 | 4 - 5 | 742 | 59 | |
XM_018057639.1 | XP_017913128.1 | 4 - 5 | 704 | 59 | ||
XM_018057648.1 | XP_017913137.1 | 4 - 5 | 835 | 59 | ||
XM_018057654.1 | XP_017913143.1 | 4 - 5 | 821 | 59 | ||
GenBank mRNA | JO410647.2 | - | 4 - 5 | 743 | 59 | |
JO425917.1 | - | 4 - 5 | 777 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |