Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAREM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018039594.1 | XP_017895083.1 | 5 - 6 | 1842 | 69 | |
GenBank mRNA | KA325078.1 | - | 5 - 6 | 1806 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Capra hircus Probe spans exons |