Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGLYRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005682223.3 | XP_005682280.2 | 5 - 6 | 1858 | 76 | |
XM_013965046.2 | XP_013820500.2 | 4 - 5 | 2005 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |