Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005684000.3 | XP_005684057.1 | 4 - 5 | 608 | 75 | |
GenBank mRNA | JO388148.2 | - | 4 - 5 | 562 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |