Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689510.3 | XP_005689567.2 | 1 - 2 | 248 | 63 | |
GenBank mRNA | JO581803.1 | - | 1 - 2 | 251 | 63 | |
JT707995.1 | - | 1 - 2 | 100 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |