Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005691626.3 | XP_005691683.2 | 2 - 3 | 219 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Capra hircus Probe spans exons |