Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042167.1 | XP_017897656.1 | 13 - 14 | 1961 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |