Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102183441 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059977.1 | XP_017915466.1 | 3 - 4 | 741 | 57 | |
XM_018059978.1 | XP_017915467.1 | 4 - 5 | 642 | 57 |
Gene Symbol: LOC102183990 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059979.1 | XP_017915468.1 | 3 - 4 | 819 | 57 | |
XM_018059980.1 | XP_017915469.1 | 4 - 5 | 639 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Capra hircus Probe spans exons |