Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCNN1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018060547.1 | XP_017916036.1 | 2 - 3 | 483 | 80 | |
GenBank mRNA | KA315733.1 | - | 1 - 2 | 157 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |