Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCNN1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018060547.1 | XP_017916036.1 | 8 - 9 | 1343 | 78 | |
XM_018060548.1 | XP_017916037.1 | 7 - 8 | 963 | 78 | ||
GenBank mRNA | KA315733.1 | - | 7 - 8 | 1017 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |