Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061146.1 | XP_017916635.1 | 7 - 8 | 4029 | 81 | |
XM_018061151.1 | XP_017916640.1 | 8 - 9 | 4052 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |