Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTUB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018066511.1 | XP_017922000.1 | 2 - 3 | 306 | 70 | |
GenBank mRNA | KA332420.1 | - | 4 - 5 | 3386 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |