Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGF29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040164.1 | XP_017895653.1 | 5 - 6 | 495 | 98 | |
XM_018040165.1 | XP_017895654.1 | 6 - 7 | 535 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |