Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKRIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040757.1 | XP_017896246.1 | 3 - 4 | 358 | 65 | |
GenBank mRNA | JO597839.2 | - | 3 - 4 | 300 | 65 | |
JO597841.2 | - | 3 - 4 | 300 | 65 | ||
KA308906.1 | - | 3 - 4 | 887 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |