Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HELT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041818.1 | XP_017897307.1 | 3 - 4 | 308 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Capra hircus Probe spans exons |