Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102183400 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018044110.1 | XP_017899599.1 | 11 - 12 | 1107 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |