Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EVX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018047522.1 | XP_017903011.1 | 1 - 2 | 653 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |