Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADRA1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018052356.1 | XP_017907845.1 | 1 - 2 | 1330 | 74 | |
XM_018052358.1 | XP_017907847.1 | 1 - 2 | 1330 | 74 | ||
XM_018052359.1 | XP_017907848.1 | 1 - 2 | 1330 | 74 | ||
GenBank mRNA | JO413605.2 | - | 3 - 4 | 1319 | 74 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Recommended Assay Capra hircus Probe spans exons |