Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRADC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005686372.3 | XP_005686429.1 | 3 - 4 | 386 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |