Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005687443.3 | XP_005687500.1 | 2 - 3 | 1154 | 76 | |
GenBank mRNA | JO616285.2 | - | 1 - 2 | 319 | 76 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Recommended Assay Capra hircus Probe spans exons |