Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689544.3 | XP_005689601.2 | 10 - 11 | 1442 | 87 | |
GenBank mRNA | JO640525.2 | - | 6 - 7 | 769 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |