Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005690886.3 | XP_005690943.1 | 2 - 3 | 426 | 72 | |
GenBank mRNA | JO439929.2 | - | 553 | 72 | ||
JO615803.2 | - | 4 - 5 | 567 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |