Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693447.2 | XP_005693504.2 | 4 - 5 | 356 | 60 | |
XM_005693448.2 | XP_005693505.2 | 4 - 5 | 592 | 60 | ||
XM_005693449.3 | XP_005693506.2 | 4 - 5 | 1521 | 60 | ||
XM_018064532.1 | XP_017920021.1 | 4 - 5 | 432 | 60 | ||
XM_018064533.1 | XP_017920022.1 | 4 - 5 | 432 | 60 | ||
GenBank mRNA | KA335455.1 | - | 3 - 4 | 274 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |