Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C19H17orf49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064536.1 | XP_017920025.1 | 5 - 6 | 745 | 63 | |
XM_018064538.1 | XP_017920027.1 | 4 - 5 | 643 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Capra hircus Probe spans exons |