Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695358.3 | XP_005695415.1 | 4 - 5 | 1156 | 58 | |
GenBank mRNA | JO420343.2 | - | 1 - 2 | 81 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |