Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013973778.2 | XP_013829232.1 | 2 - 3 | 288 | 82 | |
GenBank mRNA | JI861653.1 | - | 1 - 2 | 170 | 82 | |
JO320350.2 | - | 2 - 3 | 262 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |