Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMRN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042186.1 | XP_017897675.1 | 3 - 4 | 470 | 68 | |
GenBank mRNA | JO415925.2 | - | 3 - 4 | 473 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |