Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043633.1 | XP_017899122.1 | 3 - 4 | 792 | 62 | |
GenBank mRNA | JO343269.2 | - | 1 - 2 | 112 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Probe spans exons |