Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NACAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018047040.1 | XP_017902529.1 | 4 - 5 | 6801 | 67 | |
XM_018047041.1 | XP_017902530.1 | 4 - 5 | 6789 | 67 | ||
XM_018047042.1 | XP_017902531.1 | 5 - 6 | 5950 | 67 | ||
GenBank mRNA | JT665620.1 | - | 3 - 4 | 240 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Probe spans exons |