Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102181119 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013965615.2 | XP_013821069.2 | 1 - 2 | 3179 | 88 | |
XM_018050260.1 | XP_017905749.1 | 1 - 2 | 3179 | 88 | ||
GenBank mRNA | JT714698.1 | - | 3 - 4 | 2835 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |