Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102169288 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051811.1 | XP_017907300.1 | 4 - 5 | 583 | 136 | |
GenBank mRNA | JO598774.2 | - | 4 - 5 | 710 | 136 | |
KA311590.1 | - | 2 - 3 | 245 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |