Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102169288 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051811.1 | XP_017907300.1 | 5 - 6 | 737 | 62 | |
GenBank mRNA | JO598774.2 | - | 3 - 4 | 556 | 62 | |
KA311590.1 | - | 3 - 4 | 399 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |