Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018057758.1 | XP_017913247.1 | 2 - 3 | 364 | 80 | |
GenBank mRNA | JO626421.2 | - | 2 - 3 | 333 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |