Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018057758.1 | XP_017913247.1 | 6 - 7 | 1163 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |