Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGTRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018060339.1 | XP_017915828.1 | 2 - 3 | 164 | 79 | |
XM_018060340.1 | XP_017915829.1 | 2 - 3 | 164 | 79 | ||
GenBank mRNA | JO428138.1 | - | 3 - 4 | 2389 | 79 | |
JO428142.1 | - | 3 - 4 | 2388 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |