Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061669.1 | XP_017917158.1 | 4 - 5 | 866 | 60 | |
XM_018061670.1 | XP_017917159.1 | 4 - 5 | 778 | 60 | ||
XM_018061671.1 | XP_017917160.1 | 4 - 5 | 700 | 60 | ||
XM_018061672.1 | XP_017917161.1 | 3 - 4 | 790 | 60 | ||
XM_018061673.1 | XP_017917162.1 | 3 - 4 | 706 | 60 | ||
XM_018061674.1 | XP_017917163.1 | 3 - 4 | 555 | 60 | ||
XM_018061675.1 | XP_017917164.1 | 4 - 5 | 866 | 60 | ||
GenBank mRNA | KA341371.1 | - | 4 - 5 | 837 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |