Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDLRAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678379.3 | XP_005678436.1 | 2 - 3 | 472 | 111 | |
XM_005678380.3 | XP_005678437.1 | 4 - 5 | 442 | 111 | ||
XM_005678381.3 | XP_005678438.1 | 3 - 4 | 313 | 111 | ||
XM_013962818.2 | XP_013818272.1 | 3 - 4 | 282 | 111 | ||
XM_018044441.1 | XP_017899930.1 | 1 - 2 | 3240 | 111 | ||
GenBank mRNA | JO509608.2 | - | 2 - 3 | 653 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Capra hircus Probe spans exons |