Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042677.1 | XP_017898166.1 | 3 - 4 | 1053 | 82 | |
GenBank mRNA | JO432770.2 | - | 1 - 2 | 986 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |