Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005681639.3 | XP_005681696.1 | 2 - 3 | 332 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |