Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005681753.2 | XP_005681810.1 | 2 - 3 | 195 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |