Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055955.1 | XP_017911444.1 | 2 - 3 | 650 | 77 | |
GenBank mRNA | JO346250.2 | - | 2 - 3 | 253 | 77 | |
JT680803.1 | - | 2 - 3 | 279 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |