Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AADACL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005690882.3 | XP_005690939.2 | 4 - 5 | 497 | 86 | |
XM_013970413.2 | XP_013825867.2 | 3 - 4 | 593 | 86 | ||
GenBank mRNA | KA313053.1 | - | 1 - 2 | 1841 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |