Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CINP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005695419.3 | XP_005695476.2 | 2 - 3 | 226 | 90 | |
XM_018066022.1 | XP_017921511.1 | 2 - 3 | 226 | 90 | ||
XM_018066023.1 | XP_017921512.1 | 2 - 3 | 226 | 90 | ||
GenBank mRNA | JO402024.2 | - | 3 - 4 | 731 | 90 | |
JO443182.2 | - | 2 - 3 | 231 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |