Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018039325.1 | XP_017894814.1 | 4 - 5 | 628 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |