Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35F4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018053799.1 | XP_017909288.1 | 6 - 7 | 1169 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |