Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIF1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055992.1 | XP_017911481.1 | 4 - 5 | 327 | 72 | |
XM_018055993.1 | XP_017911482.1 | 3 - 4 | 259 | 72 | ||
XM_018055994.1 | XP_017911483.1 | 3 - 4 | 190 | 72 | ||
GenBank mRNA | JO555811.2 | - | 4 - 5 | 244 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |