Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059314.1 | XP_017914803.1 | 2 - 3 | 259 | 102 | |
XM_018059315.1 | XP_017914804.1 | 2 - 3 | 3300 | 102 | ||
XM_018059316.1 | XP_017914805.1 | 2 - 3 | 441 | 102 | ||
XM_018059317.1 | XP_017914806.1 | 3 - 4 | 3285 | 102 | ||
XM_018059318.1 | XP_017914807.1 | 3 - 4 | 3203 | 102 | ||
GenBank mRNA | JO398411.2 | - | 2 - 3 | 133 | 102 | |
KA308783.1 | - | 2 - 3 | 258 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Capra hircus Probe spans exons |