Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041076.1 | XP_017896565.1 | 3 - 4 | 311 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Probe spans exons |