Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL18RAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054705.1 | XP_017910194.1 | 1 - 2 | 75 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |