Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5DC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054714.1 | XP_017910203.1 | 2 - 3 | 150 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |